Learn More
Description
- Use qualitative technique for differential staining of bacteria from cultures and patient specimens by modified Hucker Gram stain method
- Ideal for use with Midas III and other automated staining systems
Order Info
Individual reagents supplied in packs of four 500mL bottles
Specifications
Specifications
| Type | Crystal Violet |
| Quantity | 4 x 500 mL |
| Format | Bottle |
| For Use With (Application) | Gram staining of bacteria |
| Stain Type | Crystal Violet |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.