Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ Hemacolor™ Solution II

Catalog No. 23750014 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-750014 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-750014 Supplier MilliporeSigma™ Supplier No. 65044B85
This item is not returnable. View return policy

Replacement for eosin component of the Hemacolor™ Stain Set

TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 L
Boiling Point 99.9°C
Format Liquid
Type Hemacolor Solution II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.