Learn More
MilliporeSigma™ Harleco™ Histology/Papanicolaou Stains: Gomori Trichrome
Catalog No. 23749987 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
23-749987 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-749987 Supplier MilliporeSigma™ Supplier No. 62371
This item is not returnable. View return policy
Harleco™ Histology/Papanicolaou Stains
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Connective tissue stain for collagen and muscle tissue
Quantity 500 mL
Product Type Gomori Trichrome