Learn More
MilliporeSigma™ Harleco™ Histology/Papanicolaou Stains: Hematoxylin
Catalog No. 23750019 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1000mL
4000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-750019 4000mL
23-750018 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-750019 Supplier MilliporeSigma™ Supplier No. 6506785
This item is not returnable. View return policy
Harleco™ Histology/Papanicolaou Stains
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For routine tissue specimens glycol and methylmethacrylate sections, for regressive staining
Quantity 4000mL
Product Type Gill 3 Hematoxylin