Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HARLECO™ Schiff Reagent Hotchkiss McManus

Catalog No. 23749982 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-749982 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-749982 Supplier MilliporeSigma™ Supplier No. 607371
This item is not returnable. View return policy

For Periodic acid-Schiff reaction

  • Used as replacement for Schiff Reagent in PAS kit

TRUSTED_SUSTAINABILITY

Specifications

Format Bottle
For Use With (Application) Used as replacement for Schiff's reagent in PAS kit
Packaging Glass Bottle
Quantity 500 mL
Product Type Schiff Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.