Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ HDAC1α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17608MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-608-MI 10 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-608-MI Supplier MilliporeSigma™ Supplier No. 17608
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects HDAC1α Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC1α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-HDAC1 (mouse monoclonal IgG1, supernatant). One vial containing 40μL of culture supernatant with 0.05% sodium azide. Negative ChIP Control Supernatant. One vial containing 40μL of mouse IgG containing supernatant with 0.05% sodium azide. ChIP Primers p21. One vial containing 75μL of 5μM of each primer specific for a region of the human p21 (WAF1/CIP1/CDKN1A) promoter. P21 Forward: CCC ACA GCA GAG GAG AAA GAA; P21 Reverse: CTG GAA ATC TCT GCC CAG ACA
Gene Accession No. Q13547
Gene Symbols HD1, HDAC1, RPD3L1
Host Species Mouse
Immunogen The HDAC1 antibody is made against a peptide corresponding to amino acids 467-482 of mouse HDAC1.
Quantity 10 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_032254
Test Specificity HDAC1
Includes This ChIPAb+ HDAC1 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage −20°C for one year from date of shipment
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.