Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ HDAC2α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710237MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-37MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-37MI Supplier MilliporeSigma™ Supplier No. 1710237
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects HDAC2α Clone: in Bovine, Hamster, Human, Mouse, Rat samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly conserved from yeast to human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HDAC2α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-HDAC2 (mouse monoclonal). One vial containing 50μg of purified mouse monoclonal IgG in buffer containing 70% storage buffer (0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide) and 30% glycerol. Concentration: 0.94mg/mL. Normal Mouse IgG. One vial containing 125μg of purified mouse IgG in 125μL of storage buffer containing 0.1% sodium azide. ChIP Primers, VWF promoter. One vial containing 75μL of 5μM of each primer specific for the promoter region of human von Willebrand factor. FOR: GCT GAG AGC ATG GCC TAG GGT GGT GGG CGG CAC; REV: CCC CTG CAA ATG AGG GCT GCG GCT ATC TCC AAG
Gene Accession No. Q92769
Gene Symbols HDAC2, HD2, YAF1, RPD3
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide (CEKTDTKGTKSEQLSNP) corresponding to human HDAC2 at the C-terminus.
Purification Method Protein G purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001518
Test Specificity This antibody recognizes HDAC2 at the C-terminus.
Includes This ChIPAb+ HDAC2 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Bovine, Hamster, Human, Mouse, Rat
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.