Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hematoxylin cryst. Dye

Catalog No. M1043020100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1043020100 100g
M1043020025 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1043020100 Supplier MilliporeSigma™ Supplier No. 1043020100
Add to Cart
Add to Cart

Hematoxylin cryst. (C.I. 75290) for microscopy. CAS 517-28-2, molar mass 302.28g/mol.

Hematoxylin cryst. is a dye for the specific staining of DNA in the nuclei. It is used in cytology and histology. Nuclei are stained blue, dark violet to black. Hematoxylins are mixed with trivalent positively charged metal salts and build up the so-called hematoxylin lakes, used for the selective staining of nuclei (DNA). Certistain™ is the product group for seleceted high quality dyes that fulfill all quality requirements for IVD. It is also CE certified.
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
For Use With (Equipment) Staining Equipment
Product Type Hematoxylin
Quantity 100g
Solution Type Stain
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.