Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Histone Deacetylase 6, GST-Fusion, Human Recombinant Protein, S. frugiperda

Catalog No. 38218050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-218-050UG 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-218-050UG Supplier MilliporeSigma™ Supplier No. 38218050UG
Add to Cart
Add to Cart
Full length, recombinant, human histone deacetylase 6 (HDAC6) fused to GST at the N-terminus and expressed in S. frugiperda insect cells using a baculovirus expression system. Actively maintained in the cytoplasm, HDAC6 is uniquely dependent on its ubiquitin-binding domain and functions at the nexus between two cellular signaling systems, involving protein acetylation and ubiquitination. The active deacetylase is useful in the study of HDAC6 regulation and for inhibitor screening.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 138mM NaCl, 25mM Tris-HCl, 0.05% Tween™-20 detergent, 10% glycerol, pH 8.0.
Purification Method SDS-PAGE
Quantity 50μg
Source S. frugiperda
Immunogen Histone Deacetylase 6
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.