Learn More
MilliporeSigma™ Upstate™ Histone H2A.Bbd, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set
Shop All MilliporeSigma ProductsDescription
Specifically detects Histone H2A.Bbd Clone: in Human, Mouse samples, and it is validated for ChIP, Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | Histone H2A.Bbd |
| Applications | ChIP Assay, Electromobility Shift Assay, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Anti-Histone H2A.Bbd (rabbit polyclonal). One vial containing 100μg of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before the addition of 30% glycerol. Concentration: 0.7mg/mL . Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide.ChIP primers, PGK1. One vial containing 75μL of 5μM of each primer specific for human PGK1. FOR: CCT TCC CCC ACG CTC CTA; REV: TTG TGC AGG AAC CTG TGA GAT G |
| Gene Accession No. | P0C5Y9 |
| Gene Symbols | H2A.Bbd, H2AFB1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to human Histone H2A.Bbd. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.