Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H2A.Z, Sheep Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710048MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-48MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-48MI Supplier MilliporeSigma™ Supplier No. 1710048

Sheep Polyclonal Antibody

Specifically detects Histone H2A.Z Clone: in Human samples, and it is validated for ChIP, Western Blotting

H2A.Z, also known as H2AFZ, is a variant of histone H2A variant that is highly conserved from yeast to humans. Like other histone variants, H2A.Z is encoded by a unique gene, resulting in a slightly divergent type of H2A with specialized functions. H2A.Z is involved in chromatin compaction by stabilizing the histone octamer within the nucleosome. It promotes gene expression through the creation of an open domain of chromatin that is more easily transcribed. H2A.Z is nonrandomly distributed throughout the genome, targeted primarily to pericentric heterochromatin, but excluded from the inactive X chromosome and the nucleolus. H2A.Z is associated with both euchromatin and facultative heterochromatin. DNA methylation can influence chromatin structure and effect gene silencing by excluding H2A.Z, and likewise, H2A.Z protects genes from DNA methylation. Thus, H2A.Z plays a role in regulating heterochromatin silencing and is involved in transcriptional control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.Z
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Histone H2A.Z (Sheep polyclonal IgG). One vial containing 25μg of affinity purified sheep polyclonal IgG in PBS, pH 7.4 containing 1% BSA with 0.02% sodium azide, before the addition of 30% glycerol. Normal Sheep IgG. One vial containing 25μg of protein G purified sheep IgG in 0.02 M potassium phosphate, 0.15M NaCl, pH 7.2, with 0.01% sodium azide, before the addition of 30% glycerol. Control Primers. One vial containing 75μL of 5μM of each primer specific for human GAPDH. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. P0C0S5
Gene Symbols H2A.z, H2A/z , H2AZ, MGC117173, H2AFZ
Host Species Sheep
Immunogen Linear peptide corresponding the C-terminal amino acids 113-127 of human Histone H2A.Z.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_036544
Test Specificity Recognizes Histone H2A.Z, Mr ∽15kDa.
Includes This ChIPAb+ Histone H2A.Z -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.