Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H2B, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710054MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-54MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-54MI Supplier MilliporeSigma™ Supplier No. 1710054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Histone H2B Clone: in Chicken, Human, S. cerevisiae, Xenopus, Yeast samples, and it is validated for Chromatin Immunoprecipitation, Western Blotting

Histone H2B is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the 'beads on a string' structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Histone H2B (rabbit polyclonal). One vial containing 25μL of protein A purified rabbit polyclonal in buffer containing 0.02 M phosphate buffer, pH 7.6, 0.25 M NaCl, and 0.1% sodium azide, and 30% glycerol. Normal Rabbit IgG. One vial containing 125μg of purified rabbit IgG in 125μL storage buffer. ChIP Primers, β-globin. One vial containing 75μL of 5μM of each primer specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q8N257
Gene Symbols GL105, H2B, H2B-GL105, H2B.1, H2B.q, H2B/q, H2BFQ, MGC119802, MGC119804, MGC129733, MGC129734, OTTHUMP00000013920
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 118-126 (CG-AVTKYTSSK) of human Histone H2B, with an N-terminal CG added for conjugation purposes.
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_778225
Test Specificity Recognizes and is specific for Histone H2B, MW ∽15kDa. An additional band at ∽23kDa is detected in some samples, which likely corresponds to ubiquityl-Histone H2B.
Includes The ChIPAb+ Histone H2B set includes the Histone H2B antibody, a negative control rabbit IgG and qPCR primers which amplify a 110bp region of human β-globin promoter.
Target Species Chicken, Human, S. cerevisiae, Xenopus species, Yeast
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.