Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H3.3, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710245MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-45MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-45MI Supplier MilliporeSigma™ Supplier No. 1710245
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Histone H3.3 Clone: in Human, Mouse samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone variant H3.3 is typically enriched in active chromatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.3
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Histone H3.3 (rabbit polyclonal). One vial containing 50μg of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. Control Primers, GAPDH Coding Region D2 Human. One vial containing 75μL of 5μM of each primer specific for human GAPDH coding region. FOR: GCC ATG TAG ACC CCT TGA AGA G; REV: ACT GGT TGA GCA CAG GGT ACT TTA T
Gene Accession No. P84243
Gene Symbols H3F3A, H3.3A, H3F3, PP781, H3F3B, H3.3B
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3.3.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_002098
Test Specificity This antibody recognizes human Histone H3.
Includes This ChIPAb+ Histone H3.3 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.