Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H3 (CT), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-54MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-54MI Supplier MilliporeSigma™ Supplier No. 1710254
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects Histone H3 (CT) Clone: in Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast samples, and it is validated for ChIP, Dot Blot, Immunocytochemistry, Peptide Inhibition Assay, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3 (CT)
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Histone H3 (C-Term) (rabbit polyclonal). One vial containing 25μL of rabbit antiserum diluted 1:2 in storage buffer (0.02M Phosphate, 0.25M NaCl, 0.1% sodium azide) before the addition of glycerol to 30%. Normal Rabbit Serum. One vial containing 25μL of antiserum containing 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of 5μM of each primer specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q16695
Gene Symbols HIST3H3, H3FT, MGC126886, H3t, MGC126888, H3T, H3/g, H3.4, H3/t
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_002107.3
Test Specificity Recognizes C-Terminal region of Histone H3, Mr 17kDa
Includes This ChIPAb+ Histone H3 (C-Term) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.