Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H3 (CT)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710046MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-46MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-46MI Supplier MilliporeSigma™ Supplier No. 1710046
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects Histone H3 (CT)α Clone: in Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast samples, and it is validated for ChIP, Dot Blot, Immunoprecipitation, Western Blotting

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Histone proteins are highly post-translationally modified, and Histone H3 is the most extensively modified of the five histones. The term Histone H3 alone is purposely ambiguous in that it does not distinguish between sequence variants or modification state. Histone H3 is an important protein in the emerging field of epigenetics, where its sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3 (CT)α
Applications ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Histone H3 (C-term) (rabbit monoclonal IgG). One vial containing 100μL of cultured supernatant in 0.05% sodium azide. Negative Control Supernatant (rabbit). One vial containing 100μL of cultured supernatant in 0.05% sodium azide. ChIP Primers, β-Globin. One vial containing 75μL of 5μM of each primer specific for human β-Globin. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q16695
Gene Symbols H3.4, H3/g, H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corres-ponding to the C-terminus of human Histone H3.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003484
Test Specificity Recognizes Histone H3 near the C-terminus.
Includes This ChIPAb+ Histone H3 (C-term) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Culture Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.