Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ Histone H4α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710047MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-100-47MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-47MI Supplier MilliporeSigma™ Supplier No. 1710047
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Specifically detects Histone H4α Clone: in Human samples, and it is validated for ChIP, Western Blotting

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the 'beads on a string' structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H4α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Histone H4 (rabbit monoclonal IgG). One vial containing 50μL of protein A purified rabbit monoclonal IgG supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before addition of glycerol to 40%. Normal Rabbit IgG. One vial containing 75μg of normal rabbit IgG in 75μL storage buffer. ChIP Primers, β-Globin. One vial containing 75μL of 5μM of each primer specific for human β-Globin. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corres-ponding to amino acids 17-28 of Histone H4.
Purification Method Protein A purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001029249
Test Specificity Recognizes Histone H4.
Includes This ChIPAb+ Histone H4 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.