Learn More
MilliporeSigma™ Hose Connector 1/4 in. NPTF quick-release nipple
Catalog No. XX6700028 Shop All MilliporeSigma Products
Change view
Click to view available options
Diameter (English) Outside:
1/4 in.
Catalog No. Diameter (English) Outside
XX6700028 1/4 in.
1 options
Catalog No. XX6700028 Supplier MilliporeSigma™ Supplier No. XX6700028
Add to Cart
Add to Cart
Accessories for filter holders for sample preparation.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outside 1/4 in.
Diameter (Metric) Outside 6.3 mm
Includes Hose Connector 1/4 in. NPTF quick-release nipple
Material Stainless Steel
Sterility Non-sterile
Type Laboratory Filter Holders or Cyclones
Maximum Pressure 9 bar (125 psi)
Case Material Series 300 stainless steel