Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ HP1γα, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17646MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-646-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-646-MI Supplier MilliporeSigma™ Supplier No. 17646

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects HP1γα Clone: in Human, Mouse, Vertebrate samples, and it is validated for Chromatin Immunoprecipitation, Functional Assay, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen HP1γα
Applications ChIP Assay, Functional Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation HP1γ (mouse monoclonal IgG). One vial containing 125μL of ascites. Normal Mouse IgG. One vial containing 125μg purified mouse IgG in 125μL storage buffer containing 0.1% sodium azide. ChIP Primers GAPDH Coding Region. One vial containing 75μL of 5μM of each primer specific for the coding region of human GAPDH. FOR: GGC TCC CAC CTT TCT CAT CC; REV: GGC CAT CCA CAG TCT TCT GG
Gene Accession No. Q13185
Gene Symbols CBX3; HECH; HP1-GAMMA; HP1Hs-gamma
Host Species Mouse
Immunogen HP1γ ascites is made against a GST fusion protein corresponding to amino acids 17-173 of human HP1γ.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_009207
Test Specificity Recognizes HP1γ, MW: ∽25kDa.
Includes This ChIPAb+ HP1γ -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse, Vertebrates
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.