Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HPTLC Cellulose Sheets, Aluminum Backed

Catalog No. M1160920001 Shop All MilliporeSigma Products
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
M1160920001 20 x 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1160920001 Supplier MilliporeSigma™ Supplier No. 1160920001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

used to analyze polar substances

Cellulose plates are used to analyze polar substances. An organic sorbent, cellulose is perfect for separating hydrophilic substances by partition chromatography. Typical applications include the analysis of amino acids, carbohydrates, and phosphates as well as nucleic acid and nucleic acid derivatives. We offer cellulose plates in two grades: TLC for conventional and HPTLC for demanding, high-performance separations. aluminum HPTLC Cellulose plates. Cellulose HPTLC plates size 20 x 20 cm, 25 sheets.
TRUSTED_SUSTAINABILITY

Specifications

Backing Material Aluminum
Material Cellulose
Pore Size 60 Å
Quantity 25/Pk.
Type Cellulose HPTLC Plate
Fluorescent Indicator No
For Use With (Application) Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective
Sorbent Cellulose
Thickness (Metric) 200 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.