Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hsp90, Human, Protein

Catalog No. 38589850UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-589-850UG 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-589-850UG Supplier MilliporeSigma™ Supplier No. 38589850UG
Add to Cart
Add to Cart
Native, human Hsp90. A ∽90kDa heat shock protein that is constitutively expressed and resides in the cytoplasm. It exists as a dimer and binds to other cellular proteins such as p60v-src, steroid receptors, heme-regulated protein kinase, actin, and tubulin. Binds to histones with high affinity and induces chromatin condensation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM Tris-HCl, 1mM DTT, pH 7.5.
Purification Method SDS-PAGE
Quantity 50μg
Immunogen Hsp90
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Native
Form Liquid
Purity or Quality Grade ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.