missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human APRIL Recombinant Protein

Catalog No. GF177 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF177 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF177 Supplier MilliporeSigma™ Supplier No. GF177
Only null left
Add to cart
Add to cart

Specifications

Name Human APRIL Recombinant Protein
Quantity 10 μg
Source E. coli
Recombinant Recombinant
Purity or Quality Grade Endotoxin level is less than 0.1ng/μg of protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.