Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human BMP-2 Recombinant Protein, Animal Free

Catalog No. GF319 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF319 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF319 Supplier MilliporeSigma™ Supplier No. GF319
Add to Cart
Add to Cart

Specifications

Name Human BMP-2 Recombinant Protein, Animal Free
Quantity 10 μg
Source E. coli
Recombinant Recombinant
Purity or Quality Grade Endotoxin level is less than 0.1ng/μg of protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.