Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human Mesenchymal Stem Cells (Derived from hES Cells)

Catalog No. SCC036 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC036 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC036 Supplier MilliporeSigma™ Supplier No. SCC036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes 1 million viable cells of Human Mesenchymal Stem Cells (derived from hES cells).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 kit containing 1 million viable cells
For Use With (Application) Stem cell culture
Quantity 1 Kit
Source Human ES Cells (Mel-1 cell line)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.