Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Human Recombinant MeCP2 (methyl-CpG-binding protein 2)

Catalog No. 141067 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-106-7 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-106-7 Supplier MilliporeSigma™ Supplier No. 141067
Only null left
Add to Cart
Add to Cart

Specifications

Characterization >95% purity by SDS PAGE
Concentration 0.5μg/μL
For Use With (Application) DNA-binding Proteins
Formulation In buffer containing 20mM Tris-Cl pH 7.6, 1mM EDTA, 0.15M NaCl, 10% glycerol, 0.5mM PMSF and 1mM DTT.
Name MeCP2 (methyl-CpG-binding protein 2)
Purification Method Fast Protein Liquid Chromatography
Quantity 10 μg
Source SF9 insect cells
Storage Requirements -80°C for 6 months
Conjugate N-terminal His-Tag
Cross Reactivity Mouse
Recombinant Recombinant
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.