Learn More
MilliporeSigma™ HumanKine™ Human IL-23 Recombinant Protein, Xeno-Free
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Name | Human IL-23 Recombinant Protein, Xeno-Free |
| Quantity | 10 μg |
| Source | Human 293 protein expression system |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin free and greater than 95% pure by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.