Learn More
Description
Specifications
Specifications
| Concentration | Concentration on Label |
| Formulation | Sterile-filtered (0.22μM) in 40mM Tris-HCl, pH 7.4, carrier-free. |
| Quantity | 100μg |
| Source | E. coli |
| Immunogen | γ-Interferon |
| Storage Requirements | Following initial thaw, aliquot and freeze (−20°C). Do not store in glass vials without addition of carrier protein. Further dilute in low endotoxin medium or buffer containing FBS or tissue culture grade BSA. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.