Learn More
MilliporeSigma™ γ-Interferon, Mouse, Lyophilized, Recombinant Protein, E. coli
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized from 0.5X PBS, pH 7.2. |
| Preparation Method | Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use. |
| Purification Method | RP-HPLC, FPLC, and SDS-PAGE |
| Quantity | 100 μg |
| Source | E. coli |
| Immunogen | γ-Interferon |
| Storage Requirements | Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for 2 to 7 days at 4°C or for up to 4 months at −20°C. Avoid freeze/thaw cycles of solutions. |
| Regulatory Status | RUO |
| Species | Mouse |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.