Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ γ-Interferon, Mouse, Recombinant Protein, E. coli

Catalog No. 40730320UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-730-320UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-730-320UG Supplier MilliporeSigma™ Supplier No. 40730320UG
Add to Cart
Add to Cart
Recombinant, mouse γ-interferon, consisting of 134 amino acids, expressed in E. coli as a single, non-glycosylated polypeptide. A multi-functional protein exhibiting antiviral and antitumor properties. Regulates the development of specific immune responses. Protects against bacterial sepsis in murine models.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 10mM PBS, pH 7.4.
Preparation Method Dilute only in buffers containing protein such as 0.1% BSA.
Purification Method SDS-PAGE
Quantity 20 μg
Source E. coli
Immunogen γ-Interferon
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.