Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ α-Interferon, Mouse, Recombinant Protein, E. coli

Catalog No. 407293100KU Shop All MilliporeSigma Products
Change view
Click to view available options
100kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
40-729-3100KU 100kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-729-3100KU Supplier MilliporeSigma™ Supplier No. 407293100KU
Only null left
Add to Cart
Add to Cart
Recombinant, mouse α-interferon expressed in E. coli. Antiviral and anti-neoplastic agent. Suppresses bone marrow function and induces apoptotic cell death in non-melanoma skin cancer. IFN-α signaling is mediated by JAK1 and TYK2 tyrosine kinases. Note: 1 KU = 1000 units.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In PBS containing 0.1% BSA.
Purification Method SDS-PAGE
Quantity 100kU
Source E. coli
Immunogen α-Interferon
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.