Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-1β, Human, Recombinant Protein, E. coli

Catalog No. 4076155UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-761-55UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-761-55UG Supplier MilliporeSigma™ Supplier No. 4076155UG
Add to Cart
Add to Cart
Recombinant, human interleukin-1β expressed in E. coli. Major form of interleukin-1 secreted by monocytes and macrophages. Induces nitric oxide synthase in pancreatic and smooth muscle cells and suppresses apoptosis in rat ovarian follicles. Involved in inflammatory and immune responses and promotes wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS, 50μg BSA/μg IL-1β.
Preparation Method Sterile PBS containing ≥0.1% HSA or BSA.
Purification Method SDS-PAGE
Quantity 5 μg
Source E. coli
Immunogen Interleukin-1β
Storage Requirements Following reconstitution, aliquot and freeze (−70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.