Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-10, Human, Recombinant Protein, S. frugiperda

Catalog No. 4077005UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-770-05UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-770-05UG Supplier MilliporeSigma™ Supplier No. 4077005UG
Only null left
Add to Cart
Add to Cart
Recombinant, human interleukin-10 expressed in S. frugiperda insect cells. Pleiotropic cytokine that can exert either immunosuppressive or immunostimulatory effects on a variety of cell types. Potent modulator of monocyte/macrophage function. Exhibits anti-inflammatory effects by suppressing macrophage proliferation. Blocks lipopolysaccharide induction of MMP-2 and MMP-9. Known to reduce neuronal degeneration following CNS injury by blocking glutamate-induced caspase-3 activation. Human IL-10 is also active on murine cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Carrier-free.
Preparation Method Reconstitute in sterile water or buffer to a final concentration of 0.1-1mg/mL as stock solution. Further dilute in buffer such as PBS, 1% BSA.
Purification Method SDS-PAGE
Quantity 5 μg
Source S. frugiperda
Immunogen Interleukin-10
Storage Requirements Following reconstitution, aliquot and freeze −70°C. Stock solutions are stable for up to 3 months at −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.