Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-6, Human, Recombinant Protein, E. coli

Catalog No. 40765210UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-765-210UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-765-210UG Supplier MilliporeSigma™ Supplier No. 40765210UG
Add to Cart
Add to Cart
Recombinant, human, interleukin-6 expressed in E. coli. A prototypic pleiotrophic, pro-inflammatory cytokine that stimulates the production of acute phase proteins by hepatocytes. Also known to induce skeletal muscle protein breakdown. Induces growth and differentiation of B cells, T cells, and hepatocytes. Stimulates growth and inhibits constitutive, protein synthesis-independent apoptosis of murine B cell hybridoma 7TD1. IL-6 is produced by macrophages, endothelial cells, and activated T helper cells. Also considered as a potential marker of adipocyte differentiation. Human IL-6 is active on murine cells.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Carrier-free.
Preparation Method Reconstitute to 0.1-1.0mg/mL in 100mM acetic acid.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Interleukin-6
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 3 months at −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.