Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Interleukin-6 Receptor, Soluble, Human, Recombinant Protein, S. frugiperda

Catalog No. 40765325UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-765-325UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-765-325UG Supplier MilliporeSigma™ Supplier No. 40765325UG
Add to Cart
Add to Cart
Recombinant, human soluble interleukin-6 receptor expressed in S. frugiperda insect cells. Soluble form of the IL-6 receptor that binds IL-6 and mediates IL-6 signaling through interaction with gp130. Elevated serum levels of soluble IL-6 receptors have been shown to be associated with a number of pathological states including multiple myelomas, adult T cell leukemia, interstitial lung infection, and HIV infection.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS, 50μg BSA/μg IL-6 receptor.
Preparation Method Reconstitute to a concentration ≥100μg/mL with sterile PBS containing at least 0.1% HSA or BSA.
Purification Method SDS-PAGE
Quantity 25 μg
Source S. frugiperda
Immunogen Interleukin-6 Receptor
Storage Requirements Following reconstitution, aliquot and freeze (−70°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 month at 4°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.