Learn More
MilliporeSigma™ Introductory pETBlue-1 Perfectly Blunt Cloning Kit
Catalog No. 706333 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Reactions
Catalog No. Quantity
70-633-3 100 Reactions
1 options
Catalog No. 70-633-3 Supplier MilliporeSigma™ Supplier No. 706333
Add to Cart
Add to Cart
The Perfectly Blunt™ Cloning Kits are designed for simplified cloning of DNA generated by PCR using any type of DNA polymerase. This approach enables the use of high-fidelity proofreading enzymes for amplification, thus decreasing the probability of generating mutations in the target sequence. In addition, under many conditions blunt cloning is more efficient than T-cloning, most likely due to the observation that the efficiency of single dA addition by Taq DNA polymerase varies significantly depending on the sequence context of the DNA ends, and even the number of PCR cycles performed.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C.
Quantity 100 Reactions