Learn More
MilliporeSigma™ Introductory pSTBlue-1 AccepTor Vector Cloning Kit
Catalog No. 705943 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Reactions
Catalog No. Quantity
70-594-3 100 Reactions
1 options
Catalog No. 70-594-3 Supplier MilliporeSigma™ Supplier No. 705943
Add to Cart
Add to Cart
The AccepTor™ Vector Cloning Kits are designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases (i.e. KOD XL polymerase and native and recombinant Taq polymerases), which leave single 3'-dA overhangs on the reaction products. The AccepTor Vectors enable direct ligation by virtue of single 3'-dU overhangs that anneal efficiently with 3'-dA extensions on PCR products. The dU residues are converted to dT residues in vivo following transformation.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -70°C.
Quantity 100 Reactions