Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Introductory pSTBlue-1 Perfectly Blunt Cloning Kit

Catalog No. 701843 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-184-3 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-184-3 Supplier MilliporeSigma™ Supplier No. 701843
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Perfectly Blunt™ Cloning Kits are designed for simplified cloning of DNA generated by PCR using any type of DNA polymerase.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity 100 Reactions
For Use With (Application) Cloning of DNA generated by PCR using any type of DNA polymerase

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.