Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ JHDM1B, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710264MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-64MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-64MI Supplier MilliporeSigma™ Supplier No. 1710264
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects JHDM1B Clone: in Chimpanzee, Equine, Human, Mouse, Porcine, Rabbit, Rat samples, and it is validated for Chromatin Immunoprecipitation, Dot Blot, Western Blotting

JHDM1B (jumonji domain-containing histone demethylation protein 1B) and KDM2B. Due to the presence of a F-box motif, this protein is also known as the F-box/LRR-repeat protein 10 (FBXL 10). This protein functions as a histone demethylase for lysine 36 of histione H3 and preferentially demethylates dimethylated lysine 3 with weak or no activity for mono- and tri-methylted H3 lysine 36.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JHDM1B
Applications ChIP Assay, Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-JHDM1B (Rabbit Polyclonal). One vial containing 50μL of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4) and 150mM NaCl and 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of each primer (5μM) specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q8NHM5
Gene Symbols KDM2B, CXXC2, FBXL10, JHDM1B, PCCX2, Fbl10, FBL10
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human JHDM1B.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_115979
Includes This ChIPAb+ JHDM1B -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Chimpanzee, Equine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.