Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ JMJD1C, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-62MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-62MI Supplier MilliporeSigma™ Supplier No. 1710262
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects JMJD1C Clone: in Chimpanzee, Human, Rabbit, Rhesus Ma samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Western Blotting

JMJD1C (jumonji domain-containing protein 1C) is also known as TRIP8 and mutations in this histone demethylase have been linked to certain forms of autism. JMJD1C is also thought to work in reactivating genes in embryonic stem cells, pancreatic islet and gastric cancer. JMJD1C has also been implicated as a gene expression regulator. JMJD1C with WHISTLE have recently been shown to also play a role in mouse testis development. JMJD1C has been identified as a nuclear-receptor coactivator and is a candidate gene for autism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1C
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-JMJD1C (rabbit polyclonal). One vial containing 50μL of purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of each primer (5μM) specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q15652
Gene Symbols TRIP8, RP11-10C13.2, JMJD1C, JHDM2C
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human JMJD1C.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_116165
Includes This ChIPAb+ JMJD1C -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Chimpanzee, Human, Rabbit, Rhesus Monkey
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.