Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ JMJD6, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710263MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-63MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-63MI Supplier MilliporeSigma™ Supplier No. 1710263
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Specifically detects JMJD6 Clone: in Human samples, and it is validated for ChIP, Western Blotting

JmjC domain-containing protein 6 (JMJD6) is a dioxygenase belonging to group of proteins known as the Jmjc-domain containing proteins. This family of proteins is a group of redox enzymes specifically involved in catalyzing oxidation reactions, and are characterized by the presence of a highly conserved jumonji C (Jmjc)-domain. JMJD6 can function both as a lysyl-hydroxylase and a histone arginine demethylase. As a lysyl-hydroxylase, JMJD6 catalyzes 5-hydroxylation of target protein lysine residues. As an arginine demethylase, JMJD6 has a critical part to play in histone code by demethylating of histone H3 and histone H4. It has also been shown to mediate U2AF2/U2AF65 5-hydrozylation, thereby functioning as a regulator for RNA splicing through affecting U2AF2/U2AF65 pre-mRNA activity. JMJD6 is a critical component of embryogenesis for the differentiation of various organs, and may act as a regulator for hematopoietic regulation and macrophage cytokine response. High expression of JMJD6 has been observed in heart, kidney and skeletal muscle. Medium to low expression has been seen in brain, liver, lung, ovary, pancreas, placenta, prostate, testis, thymus, and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD6
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-JMJD6 (rabbit polyclonal). One vial containing 50μL of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl and 0.05% sodium azide before the addition of glycerol to 30%. Concentration: 0.7mg/mL. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, human β-globin. One vial containing 75μL of each primer (5μM) specific for the human β-globin promoter. FOR: AGG ACA GGT ACG GCT GTC ATC; REV: TTT ATG CCC AGC CCT GGC TC
Gene Accession No. Q6NYC1
Gene Symbols JMJD6, PTDSR1, PTDSR, PSR
Host Species Rabbit
Immunogen Linear peptide corresponding to human JMJD6.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001074930
Test Specificity This antibody recognizes JMJD6.
Includes This ChIPAb+ JMJD6 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.