Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lactate Dehydrogenase, Porcine Heart, Protein

Catalog No. 42721150KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-721-150KU 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-721-150KU Supplier MilliporeSigma™ Supplier No. 42721150KU
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native lactate dehydrogenase from porcine heart. A tetrameric enzyme that catalyzes the reversible conversion of pyruvate to lactate. The overall equilibrium of this reaction favors lactate formation. Note: 1KU = 1000 units.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Aqueous Buffers
Quantity 50 kU
Immunogen Lactate Dehydrogenase
Storage Requirements Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 24hr. at 4°C. Do not freeze solutions.
Regulatory Status RUO
Species Porcine
Recombinant Native
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.