Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ LEF1α, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17604MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-604-MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-604-MI Supplier MilliporeSigma™ Supplier No. 17604
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects LEF1α Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen LEF1α
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-LEF1 (mouse IgG1). 1 vial containing 100mg of thiophilic and size exclusion chromatography purified mouse IgG1 in 100mL of phosphate buffered saline with sodium azide. The antibody is made against amino acid residues 1-85 of human LEF1 and can recognize human and mouse LEF1. It does not cross-react with TCF-3 or TCF-4. Normal Mouse IgG. One vial containing 125μg of normal mouse IgG in 125μL volume. ChIP primers c-myc. One vial containing 75mL of 5μM of each control primer specific for human c-myc promoter. FOR: CCC AAA AAA AGG CAC GGA A; REV: TAT TGG AAA TGC GGT CAT GC
Gene Accession No. Q9UJU2
Gene Symbols LEF1, DKFZp586H0919, TCF1ALPHA, LEF-1, TCF1-alpha
Host Species Mouse
Immunogen The antibody is made against amino acid residues 1-85 of human LEF1.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_016269.2
Test Specificity Recognizes LEF1 (lymphoid enhancer-binding factor 1).
Includes This ChIPAb+ LEF1 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage −20°C for one year from date of shipment
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.