Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Leptin, Mouse, Recombinant Protein, E. coli

Catalog No. p-5365147 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-970-51MG 1 mg
42-970-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-970-51MG Supplier MilliporeSigma™ Supplier No. 4297051MG
Recombinant, mouse leptin espressed in E. coli. Native leptin is a product of the obese (ob) gene that serves as a ligand for the OB receptor (OB-R). Mice with mutations in the ob gene have been found to be obese and diabetic and to have reduced activity, metabolism and body temperature. Suppresses insulin secretion by inhibiting activities of Ca2+-dependent PKC isoforms. Reported to reduce hepatic glucose production by blocking phosphoenolpyruvate synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a sterile filtered PBS.
Preparation Method Reconstitute with 20mM Tris-HCl, pH 8.0 to a final concentration of 1mg/mL.
Purification Method SDS-PAGE
Quantity 1 mg
Source E. coli
Immunogen Leptin
Storage Requirements Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C or −70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C or −70°C. Avoid freeze/thaw cycles of solutions.
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.