Learn More
MilliporeSigma™ LiChrolut™ Extraction Unit Drying Attachment
Catalog No. M1198520001 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Drying Attachment, Complete
Catalog No. Type
M1198520001 Drying Attachment, Complete
1 options
Catalog No. M1198520001 Supplier MilliporeSigma™ Supplier No. 1.19852.0001
Add to Cart
Add to Cart
for sample preparation
All the individual steps associated with solid-phase extraction can be carried out using the LiChrolut™ extraction unit rapidly and reliability.This transparent and vacuum – suitable unit made of glass can be used to prepare up to 12 samples simultaneously.
TRUSTED_SUSTAINABILITY
Specifications
Type Drying Attachment, Complete