Learn More
MilliporeSigma™ LiChrolut™ Extraction Unit and Drying Attachment Accessories: Large Volume Capillaries
Catalog No. M1199020001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
LiChrolut Extraction Unit and Drying Attachment
Catalog No. For Use With (Equipment)
M1199020001 LiChrolut Extraction Unit and Drying Attachment
1 options
Catalog No. M1199020001 Supplier MilliporeSigma™ Supplier No. 1.19902.0001
for large-volume samples
All the individual steps associated with solid-phase extraction can be carried out using the LiChrolut™ extraction unit rapidly and reliability.This transparent and vacuum – suitable unit made of glass can be used to prepare up to 12 samples simultaneously.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LiChrolut Extraction Unit and Drying Attachment
Type Large Volume Capillaries
Includes Six pieces stainless steel, electro-polished 2.0 O.D. x 1.5 I.D. x 300 mm lg