Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LiChrosorb™ RP-18 Sorbents, 5μm

Catalog No. M1093330010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1093330010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1093330010 Supplier MilliporeSigma™ Supplier No. 1093330010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

well suited for the separation of neutral, acidic and weak basic compounds.

LiChrosorb™ RP-18 is a reliable irregular shaped silica gel carriers with reversed phase properties.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 g
Surface Area (Metric) 300 m2/g
Product Type HPLC Sorbent
Material Silica
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.