Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lipase, Chromobacterium viscosum, Protein

Catalog No. 437707100KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-770-7100KU 100 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-770-7100KU Supplier MilliporeSigma™ Supplier No. 437707100KU
Add to Cart
Add to Cart
Native lipase from Chromobacterium viscosum. Catalyzes hydrolysis of triglycerides to glycerol and fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Reconstitute in 100mM potassium phosphate buffer, pH 8.0, containing 1mg/mL BSA.
Quantity 100 kU
Source Chromobacterium viscosum
Immunogen Lipase
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Recombinant Native
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.