Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate™ LSF, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710252MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-102-52MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-52MI Supplier MilliporeSigma™ Supplier No. 1710252

Rabbit Polyclonal Antibody

Specifically detects LSF Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

Transcription factor LSF (late simian virus 40 factor) also known as transcription factor LSF, CP2, LBP-1c, and SEF is expressed in a variety of cell types including brain, ovary, kidney, thymus, spleen, liver, adrenal, heart and lung. LSF is essential for cell cycle progression at the G1/S transition after reentry into the cell cycle from G0. LSF plays an important role in DNA synthesis and cell survival. A major cellular target of LSF is the thymidylate synthase (TS) gene coding for the rate limiting enzyme in the production of dTTP. Disruption of LSF function results in apoptosis at S phase or to cell cycle arrest at the G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSF
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-LSF (rabbit polyclonal). One vial containing 50μL of purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide before the addition of glycerol to 30%. Normal Rabbit IgG. One vial containing 125μg of Rabbit IgG in 125μL of storage buffer containing 0.05% sodium azide. ChIP Primers, Mouse switch (S) regions α. One vial containing 75μL of 5μM of each primer specific for Mouse switch (S) regions α. FOR: CTA GAC CGG GCA GAC CTA GA; REV: AAA GCC CAT TCC AGC CTA GT
Gene Accession No. Q12800
Gene Symbols TFCP2, LSF, SEF
Host Species Rabbit
Immunogen Linear peptide corresponding to amino acids 310-335 of human LSF.
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005644
Includes This ChIPAb+ LSF -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.