Learn More
MilliporeSigma™ Luteinizing Hormone, Human Pituitary, Iodination Grade, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | H2O (1mg/mL) |
| Purification Method | By weight |
| Quantity | 10 μg |
| Immunogen | Luteinizing Hormone |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Native |
| Form | Solid |
| Purity or Quality Grade | ≥95%, Iodination Grade |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.