Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Magna GrIP™ HT96 Rack

Catalog No. 1710071 Shop All MilliporeSigma Products
Change view
Click to view available options
Array:
4 x 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array
17-100-71 4 x 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Strong magnets can cause serious injury to people with Pacemakers or Automatic Internal Cardiac Defibrillators. Strong magnets can attract tools and metal objects, causing pinch, cut or entrapment hazards. Keep tools, metal objects, and magnetic media away from magnets. Do not store near computers, cell phones, or other electronic equipment.

Catalog No. 17-100-71 Supplier MilliporeSigma™ Supplier No. 1710071
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strong magnets can cause serious injury to people with Pacemakers or Automatic Internal Cardiac Defibrillators. Strong magnets can attract tools and metal objects, causing pinch, cut or entrapment hazards. Keep tools, metal objects, and magnetic media away from magnets. Do not store near computers, cell phones, or other electronic equipment.

Designed for separation of magnetic particles from solutions in a 96-well plate.

  • Composed of an acrylic block (12.8 x 8.6cm) containing 24 (4 x 6) neodymium magnetic rods each having a diameter of approximately 7mm and a Gauss rating of 1200 (1.21 to 1.22 Teslas)
  • When a standard 96-well or PCR plate is fitted over the magnetic rack, each magnetic rod sits between four adjacent wells and rapidly pulls magnetic particles to the side or bottom of the wells
  • Holes in the rack are slightly angled to maximize exposure to the magnets and to ease removal of the liquid from the tube

Store at room temperature. To clean, wash the Magna GrIP™ HT96 rack in soap and water, and air dry. Avoid exposure to organic solvents including benzene, toluene, xylene, chloroform, and acetone.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color White
Material Acrylic
Positions 24
Shape Rectangular
Sterility Non-sterile
For Use With (Application) Separation of magnetic particles from solutions
For Use With (Equipment) 96-well plates
Dimensions (L x W) 12.8 x 8.6cm
Product Type Magnetic Rack
Array 4 x 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.