Learn More
MilliporeSigma™ May-Grünwald's Eosin Methylene Blue Dyes
Catalog No. M1013520100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25g
100g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1013520100 100g
M1013520025 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1013520100 Supplier MilliporeSigma™ Supplier No. 1013520100
Add to Cart
Add to Cart
for microscopy
The staining method according to May-Grünwald is one of the standard techniques in hematological diagnostic procedures. The May-Grünwald's eosin methylene blue dye is a mixture of dry dyes used for the self-preparation of the May-Grünwald's staining solution. Like Wright's eosin methylene blue and Leishman's eosin methylene blue, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The typical resulting color of cell nuclei is red to violet. As a CE-certified, IVD-registered product, it can be used in diagnostics and laboratory accreditations without constraints.
TRUSTED_SUSTAINABILITY
Specifications
Color Dark green
Packaging Glass Bottle
Quantity 100g
Product Type Eosin Methylene Blue Dye